Order Entry
United States
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (DyLight® 488)
Anti-IgG Donkey Polyclonal Antibody (DyLight® 488)
  RL610-741-002
undefined
Anti-IgG Donkey Polyclonal Antibody (DyLight® 488)
  RL610-741-002
 :  610-741-002
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Donkey
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    Heavy and Light chain
  • Cross Adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage Buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage Temperature:
    –20 °C
  • Concentration:
    1.0 mg/ml by UV absorbance at 280 nm
  • Shipping Temperature:
    Ambient
  • Immunogen:
    Mouse IgG whole molecule
  • Tested Applications:
    Fluoresence ELISA
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. No.:
    RL610-741-002
  • Supplier no.:
    610-741-002

 

 

The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Mouse