Anti-HIST2H3C Rabbit Polyclonal Antibody
Catalog # RL600401I60
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Primary
- Antigen Name:Histone Cluster 2, H3c
- Antigen Symbol:HIST2H3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Drosophila,Plant,Xenopus,Chicken,C. elegans,Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µg
- Cross Adsorption:No
- ChIP:Yes
- Antigen Synonyms:H3.3B|family 3A|H3R2me1|MGC87782|H3.3AH3F3H3F3B|histone H3.3|MGC87783|H3 histone
- Modification:acetyl-Lys4
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.05% (w/v) Sodium Azide
- Storage Temperature:–20 °C
- Concentration:1.15 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:Histone H3 [ac Lys4] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic acetylated peptide surrounding Lysine 4 of human Histone H3.2.
- Tested Applications:Dot Blot
- Purification:purified by immunoaffinity chromatography
- Cat. No.:RL600401I60
- Supplier no.:600-401-I60
Specifications
About this item
Anti-Histone H3 [ac Lys4] antibody is useful for Western Blot, Immunocytochemistry, Immunofluorescence, Chromatin Immunoprecipitation, and Dot Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~15.4
Recommended Dilutions: ChIP: 2-5µg/million cells; Immunofluorescence: 1:1000; Western Blot: 1:500; Immunochemistry: 1:1000; Other Dilution: Dot Blot 1:1000
Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, plant