Anti-Surface LipoProtein p27 antibody has been tested in Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~30.9 kDa in size corresponding to p27 by Western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:1000;
Type: Primary
Antigen: BBA060
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- RL200-401-C30
- RL200-401-C30S
- Antigen Symbol:
- BBA060
- BBA060
- Antigen Synonyms:
- Surface LipoProtein p27
- Surface LipoProtein p27
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- B. burgdorferi
- B. burgdorferi
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1194336
- 1194336
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- MBP-fusion Protein corresponding to Borrelia burgdorferi Surface LipoProtein p27 Protein.
- MBP-fusion Protein corresponding to Borrelia burgdorferi Surface LipoProtein p27 Protein.
- Format:
- Antibody fraction
- Antibody fraction
- Form:
- Lyophilized
- Liquid (sterile filtered)
- Purification:
- purified by immunoaffinity chromatography
- Protein A purified
- Concentration:
- 1.0 mg/ml by UV absorbance at 280 nm
- 1.0 mg/ml by UV absorbance at 280 nm
- Cross Adsorption:
- No
- No
- Storage Buffer:
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:
- –20 °C
- –20 °C
- Shipping Temperature:
- Ambient
- Dry Ice