Anti-ErpD Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemical
Certificates
About this item
Anti-ErpD antibody has been tested in Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect bands at ~37.5 kDa and ~65 kDa in size corresponding to ErpD/Arp37 by Western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:1000;
Type: Primary
Antigen: ErpD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi
Specifications
- Catalog No:
- RL200-401-C09
- RL200-401-C09S
- Antigen Symbol:
- ErpD
- ErpD
- Antigen Synonyms:
- ErpD/Arp37
- ErpD/Arp37
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- B. burgdorferi
- B. burgdorferi
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1194073
- 1194073
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- MBP-fusion Protein corresponding to Borrelia burgdorferi ErpD/Arp37 Protein.
- MBP-fusion Protein corresponding to Borrelia burgdorferi ErpD/Arp37 Protein.
- Format:
- Antibody fraction
- Antibody fraction
- Form:
- Lyophilized
- Liquid (sterile filtered)
- Purification:
- Protein A purified
- Protein A purified
- Concentration:
- 1.0 mg/ml by UV absorbance at 280 nm
- 1.0 mg/ml by UV absorbance at 280 nm
- Cross Adsorption:
- No
- No
- Storage Buffer:
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:
- –20 °C
- –20 °C
- Shipping Temperature:
- Ambient
- Dry Ice
Specifications
Related Information
Frequently Bought Together