Order Entry
United States
ContactUsLinkComponent
Anti-CD3, CD14, CD16, CD19, CD20, CD56 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT1, HCD14, 3G8, HIB19, 2H7, HCD56]
Anti-CD3, CD14, CD16, CD19, CD20, CD56 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT1, HCD14, 3G8, HIB19, 2H7, HCD56]
  348803-BL
 :  Biolegend
Anti-CD3, CD14, CD16, CD19, CD20, CD56 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT1, HCD14, 3G8, HIB19, 2H7, HCD56]
  348803-BL
 :  Biolegend
 :  348803

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD3, CD14, CD16, CD19, CD20, CD56
  • Clone:
    UCHT1, HCD14, 3G8, HIB19, 2H7, HCD56
  • Conjugation:
    APC (Allophycocyanin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b,
    IgG1
  • Reactivity:
    Human
  • Size:
    50 tests
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Antigen Synonyms:
    CD3
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Concentration:
    0.00
  • Shipping Temperature:
    Ambient RT
  • Purification:
    The antibodies are purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
  • Cat. No.:
    348803-BL
  • Supplier no.:
    348803

 

 

APC anti-human Lineage Cocktail (CD3/14/16/19/20/56) [UCHT1, HCD14, 3G8, HIB19, 2H7, HCD56]; Isotype: Mouse IgG1/Mouse IgG2b; Reactivity: Human; Apps: FC; Size: 50 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 20 μl per million cells or 20 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type:
Antigen: CD3, CD14, CD16, CD19, CD20, CD56
Clonality:
Clone: UCHT1, HCD14, 3G8, HIB19, 2H7, HCD56
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1/IgG2b
Reactivity: Human