ContactUsLinkComponent
Argos 100 Place LiteSafe™ Freezer Box
Argos 100 Place LiteSafe™ Freezer Box
Catalog # 490014-778
Supplier:  Antyila Scientific
Small Business Enterprise
undefined
Argos 100 Place LiteSafe™ Freezer Box
Catalog # 490014-778
Supplier:  Antyila Scientific
Supplier Number:  06755-21
Small Business Enterprise
Specifications
  • Description:
    100 Place LiteSafe™ Freezer Box
  • Dimensions:
    14×14×5.4 cm
  • Environmentally Preferable:
  • Cat. No.:
    490014-778
  • Supplier no.:
    R3121
Specifications
About this item
Ideal for light-sensitive specimens, this all black storage box is suitable for use with 0.5, 1.5, and 2.0 mL micro-tubes, as well as most other brands of cryogenic vials
The reinforced seal, triple hinges and locking clasp keeps samples secure while the box is closed. Each box features and alpha-numeric grid on the exterior and interior, stacking rings and a matte surface suitable for printing and labeling.