ContactUsLinkComponent
MBP Graduated Filtered Pipette Tips, 96 Tip Rack, 200 µl, Thermo Scientific
MBP Graduated Filtered Pipette Tips, 96 Tip Rack, 200 µl, Thermo Scientific
Catalog # 490014-498
Supplier:  Thermo Scientific
undefined
MBP Graduated Filtered Pipette Tips, 96 Tip Rack, 200 µl, Thermo Scientific
Catalog # 490014-498
Supplier:  Thermo Scientific
Supplier Number:  F108-96RS-200
Specifications
  • Volume:
    200 µl
  • Cat. No.:
    490014-498
  • Filter:
    +
  • Tip style:
    Graduated Filtered Pipette Tips
  • Supplier no.:
    F108-96RS-200
Specifications
About this item
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product.
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. First-Class Filter Tips: Specially designed for PCR work, these pipette tips prevent airborne contaminants from coming in contact with the barrel of the pipettor. Graduated Tips: Provides reference lines to allow a quick check of pipetting procedures by indicating nominal liquid volume in the tip. For use with but not limited to: Biohit Proline®, Costar® Onepette, Finnpipette®, Jencons Sealpette®, Labpette™, Oxford-Benchmate®, Pipetman®, and GeneMate®. Sterile.
A mold release agent is NOT used in the manufacturing of these pipette tips.Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals. A trace metal certificate is available upon request.Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free.) Certificate of analysis is available upon request.Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation. Certificate of irradiation available upon request.