ContactUsLinkComponent
Axygen® PlateMax® Heat Sealing Film with Paper Backing, Corning
Axygen® PlateMax® Heat Sealing Film with Paper Backing, Corning
  47734-815
 :  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
Axygen® PlateMax® Heat Sealing Film with Paper Backing, Corning
  47734-815
 :  Corning
 :  MF-300
Product available on GSA Advantage®
   Low Manufacturing Impact
  • Description:
    Heat Sealing Film with Paper Backing
  • Thickness:
    85 µm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    I-RECs, ISO 14001 - Reynosa
  • Cat. No.:
    47734-815
  • Supplier no.:
    MF-300
Axygen microplate heat sealing films are designed for applications ranging from PCR and Real-Time PCR to storage.
They can be used in a wide range of temperatures, making them suitable for nearly any application.