Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Density Meters
Accessories for Biowave Cell Density Meter CO8000
Accessories for Biowave Cell Density Meter CO8000
490005-908
:
BIOCHROM US BE
Accessories for Biowave Cell Density Meter CO8000
490005-908
:
BIOCHROM US BE
:
80-3000-57
Description:
Tube adapter for 10, 12 mm tubes
Cat. No.:
490005-908
Supplier no.:
80-3000-57
Details:
For use with Biochrom WPA CO8000 Cell Density Meter