Orders LinkContactUsLinkComponent
SmartWrite Slide Printer, Sakura Finetek
Slide Printer, 110 V, 50/60 Hz
  10062-828
 :  9021
SmartWrite Slide Printer, Sakura Finetek
  10062-828
SmartWrite Slide Printer, Sakura Finetek
SmartWrite Slide Printer, Sakura Finetek
Slide Printer, 110 V, 50/60 Hz
$18,600.00
Each
Microscope slide printer with thermal transfer print technology, with multiple color or black ribbons
Prints directly on white 3 x 1" standard or positive charged slides with smooth surfaced white frosted end and clipped corners.
SwartWrite Software (9025), PC with touch screen (9026) and 2-D bar code scanner (9024) are all sold separately.
Data interface USB 2.0. Slide cartridge capacity 100 slides with slide output tray capacity 15 slides. Print resolution 300 dpi. Printer drivers are Windows XP & Windows 7 compatible. Print speed up to 9 slides/minute using black; up to 5 slides/minute using solid colors; up to 4 slides/minute using blended colors.
Scannable 2D and 1D bar Code, alphanumeric and graphic printing capabilities.
 : Warranty is one year, parts and labor.
  • Description:
    Slide Printer
  • Electrical:
    110 V, 50/60 Hz
Accessories
  •  
    Description
    For Use With
  • 10062-830
    2-D Bar Code Scanner, Stand included
    SmartWrite Printers
    Each - $934.00
  •  
    Description
    For Use With
  • 10062-832
    SmartWrite Software, 1 License/5 Printers/1 Computer, Configurable Label Design
    SmartWrite Printers, Windows XP/7; LIS interface-NiceWatch; Runs on PC or on Network
    Each - $3,320.00
  •  
    Description
    For Use With
  • 10062-846
    SmartWrite Shuttle PC with Touch Screen and 1.3M Pixel Web Camera, IEEE 802.11 B/G/N (Complete with Laboratory Keyboard and Mouse, 2 USB Ports, 4-in-1 Card Reader, Headphone, Mic, LAN Port, D-sub Port)
    SmartWrite Printers, 1 GP PC with 250 GB SATA 5400 RPM 8 MB 2.5", Windows 7 Home Premium 32 BIT, Intel Atom D510 Dual Core CPU (1.66 GHz).
    Each - $4,300.00
  •  
    Description
    For Use With
  • 10062-848
    SmartWrite Blue Slide Holder, Holds 100 Slides
    SmartWrite Printers
    Each - $214.00
  •  
    Description
    For Use With
  • 10062-850
    SmartWrite Black Printer Ribbon; Chemical Resistant
    SmartWrite Printers
    Case of 6 - $1,110.00
  •  
    Description
    For Use With
  • 10062-840
    SmartWrite Color Printer Ribbon; Chemical Resistant
    SmartWrite Printers
    Case of 6 - $1,240.00