Specifications
- Description:Automated Slide Stainer
- Cat. No.:10062-858
- Electrical:115 V, 50/60 Hz, 1 A
- Supplier no.:1900
Specifications
About this item
Stains up to 100 slides per hour with 22 reagent reservoirs and 2 water stations
Holds up to 15 programs with up to 35 steps in each program.
Bi-directional robotic arm allows for progressive and regressive staining protocols.