ContactUsLinkComponent
Hypersil GOLD™ Phenyl HPLC Columns, Thermo Scientific
Hypersil GOLD™ Phenyl HPLC Columns, Thermo Scientific
Supplier:  Thermo Scientific
Certificates
undefined
Hypersil GOLD™ Phenyl HPLC Columns, Thermo Scientific
Supplier:  Thermo Scientific
Specifications
  • Catalog No:
  • 76332-252
  • Description:
  • 76332-252
  • Carbon Load:
  • 8.0%
  • pH Range:
  • 2 - 8
  • Surface Area:
  • 220 m²/g