ProSwift SCX -1S Protein Columns, Thermo Scientific
10044-588
- Column Format:Analytical
- Ø int.:4.60 mm
- Length:50 mm
- pH Range:2 to 12
- Description:10044-588
- Bed weight:30 mg/ml
- Cat. No.:10044-588
- Packing material:Lysozyne Sulfonic acid
- Phase:Ploymethacrylate
- Supplier no.:066765
Fast ion-exchange separation and protein purification is possible using the Thermo Scientific™ ProSwift ion exchange (IEX) columns.
- Durable and robust, these columns exhibit stability
- Reproducibility even after hundreds of runs
These monolith ion-exchange columns deliver the outstanding resolving power of nonporous media combined with the high capacity of conventional porous media. They offer excellent separations using fast gradients at low and high flow rates, which improve productivity.