ContactUsLinkComponent
ProPac™ HIC Hydrophobic Interaction Columns, Thermo Scientific
ProPac™ HIC Hydrophobic Interaction Columns, Thermo Scientific
Supplier:  Thermo Scientific
Certificates
undefined
ProPac™ HIC Hydrophobic Interaction Columns, Thermo Scientific
Supplier:  Thermo Scientific
Specifications
  • Catalog No:
  • 10040-598
  • 10044-518
  • 10044-520
  • 10044-522
  • Max. Pressure:
  • 3500 psi
  • 3500 psi
  • 3500 psi
  • 3500 psi
  • pH Range:
  • 2.5 to 7.5
  • 2.5 to 7.5
  • 2.5 to 7.5
  • 2.5 to 7.5
  • Surface Area:
  • 100 m²/g
  • 100 m²/g
  • 100 m²/g
  • 100 m²/g
  • Temperature Limit:
  • 40 °C
  • 40 °C
  • 40 °C
  • 40 °C