Acclaim™ 300 C18 Columns, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Achieve high-resolution peptide mapping and protein separations with Thermo Scientific™ Acclaim 300 C18 reversed-phase columns.
The 3 µm ultrapure silica particles reduce the limits for diffusion of the analytes into the stationary phase, resulting in fast, high-resolution separations. Compared to 5 µm column packings, the smaller particles support increased flow rates and shallower gradients on shorter columns for faster separations.
Ordering information: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (Part No. 10046-750).
Specifications
- Catalog No:
- 10044-450
- 10044-452
- 10044-454
- 10044-624
- 10044-456
- 10044-526
- 10044-638
- Description:
- 10044-450
- 10044-452
- 10044-454
- 10044-624
- 10044-456
- 10044-526
- 10044-638
- Carbon Load:
- 0.08
- 0.08
- 0.08
- 0.08
- 0.08
- 0.08
- 0.08
- pH Range:
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- Surface Area:
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
Specifications
Frequently Bought Together