Hypersil™ SAX HPLC Columns, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Highly stable silica-based quarternary amine strong anion exchange columns, designed for aqueous and low pH mobile phases.
For more challenging separations that require improved chromatographic performance, we recommend the use of Hypersil GOLD SAX columns as an alternative.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Frequently Bought Together