About this item
Nanoflow LC-MS utilizes very narrow fused silica columns and as a result, even the slightest inconsistency in column connection can result in leaks and dead volumes are a frequent source of poor data, causing unstable spray and a significant loss in performance.
Trouble-shooting connection issues can be difficult and time consuming, even for experts. Thermo Scientific EASY-Spray Columns address the aforementioned issue by incorporating high-performance nanobore columns and integrating them within a radical new design. Packed with either C18 for peptide separations or with selected wide pore phases and monoliths for the separation of intact proteins,e.g. top-down proteomics or the structural identification of biotherapeutics, the 'plug and spray' approach delivers state of the art performance in a simple to use format, making research routine.
Nano columns packed with core-enhanced Accucore C4, for top-down proteomics.
Specifications
- Catalog No:
- 76449-088
- 76449-090
- 76449-092
- 76449-082
- 76449-084
- 76449-086
- 76449-094
- 76449-096
- Description:
- 76449-088
- 76449-090
- 76449-092
- 76449-082
- 76449-084
- 76449-086
- 76449-094
- 76449-096
- Carbon Load:
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- 15%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. Pressure:
- 14500 psi
- 11600 psi
- 17400 psi
- 7250 psi
- 11600 psi
- 14500 psi
- —
- pH Range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface Area:
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- Temperature Limit:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- USP Code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1