ContactUsLinkComponent
Acclaim™ Phenyl-1 LC Columns, Thermo Scientific
Acclaim™ Phenyl-1 LC Columns, Thermo Scientific
Supplier:  Thermo Scientific
Certificates
undefined
Acclaim™ Phenyl-1 LC Columns, Thermo Scientific
Supplier:  Thermo Scientific
Specifications
  • Catalog No:
  • 10044-736
  • 10044-740
  • 10044-742
  • 10044-732
  • 10044-734
  • 10043-342
  • Description:
  • 10044-736
  • 10044-740
  • 10044-742
  • 10044-732
  • 10044-734
  • 10043-342
  • Carbon Load:
  • 13%
  • 13%
  • 13%
  • 13%
  • 13%
  • 13%
  • pH Range:
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • 2 to 8
  • Surface Area:
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g