Sonics Accessories for 50-Watt Liquid & 130-Watt Ultrasonic Processors
Supplier: SONICS & MATERIALS BE
Certificates
About this item
Optional Probes
Probes with smaller diameter produce greater intensity of cavitation, but the energy released is restricted to a more concentrated field. Probes with a larger tip diameter produce less intensity, but can process larger samples.Footswitch is ideal for hands-free operation. Includes a 10-ft cable. Use with S-2034-130 only.Converter Clamp supports a 32 mm (1.25") converter and fits a 13 mm (0.5") rod. Chemical-resistant, reinforced plastic.Sound-abating Enclosure permits extended processing without discomfort by reducing noise to a comfortable level. Support rod and converter clamp are included. Measures 12" W x 20" H x 12" D.
Frequently Bought Together