GeneMate 80-Place Microtube Racks, Scientific Specialties
Supplier: GeneMate
Certificates
About this item
These 80-Place Fraction Collection Microtube Racks are ideal for hundreds of laboratory applications
Polypropylene racks are available in assorted colors and are freezer-safe and autoclavable. Each rack features a frosted writing surface on the side of the rack and has conical shaped bottoms to keep samples centered. Racks are available in natural and assorted colors.
Frequently Bought Together