GeneMate 4-Way Flipper® Racks, Medium, Scientific Specialties
Supplier: GeneMate
Certificates
About this item
Racks will hold eight 30 mm diameter tubes (e.g., 50 mL conical tubes), twenty 20 mm diameter tubes, twenty-four 16-17 mm diameter tubes (e.g., 15 mL conical tubes), or forty 12-13 mm diameter tubes.
Frequently Bought Together