- Description:ENDURO Electroblotting System, Buffer Tank and Blotting Module
- Environmentally Preferable:
- Cat. No.:490008-972
- Supplier no.:E2010-BT
Labnet's ENDURO™ Vertical PAGE System is modular - allowing PAGE, electroblotting and 2D gel electrophoresis to be carried out in the same tank, simply by changing the inserts. Users can purchase only the parts they need, or a complete combined system for a cost savings over buying all three systems separately.
- Modular Design
- PAGE, Electroblotting and 2D Electrophoresis
- Quick and easy set up
- Wide range of accessories