About this item
- Spring-loaded clamping mechanism, alignment pins, & hollow gasketsensures leak-free PAGE gel casting
- Gels are cast directly in the Upper Buffer Chamberno need to transfer gels after polymerization
- Integrated water cooling systemprevents band distortion from excessive heat generated during electrophoresis
- Interlocking & color-coded safety lidprevents reverse electrophoresis & accidental electrical shock
- Compatible with many pre-cast gels (10 x 10 cm formats only)
Complete 10 x 10cm System Includes:
Casting Base
2 x 10-well combs, 0.8mm thick
4 side spacers
4 complete sets of glass (4 each, notched & blank)
Blocking plate
Replacement gaskets
Complete 20 x 14cm System Includes:
Casting Base
2 x 15-well combs, 2 x 20-well combs, 0.8mm thick
8 side spacers
4 complete sets of glass (4 each, notched & blank)
Blocking plate
Replacement gaskets
Complete 16 x 14cm System Includes:
Casting Base
2 x 10-well combs, 2 x 25-well combs, 1.5mm thick
8 side spacers
4 complete sets of glass (4 each, notched & blank)
Blocking plate
Replacement gaskets
Complete 20 x 20cm System Includes:
Casting Base
2 x 15-well combs, 2 x 20-well combs, 1.5mm thick
8 side spacers
4 complete sets of glass (4 each, notched & blank)
Blocking plate
Replacement gaskets