Cooling Containers
Supplier: Heathrow Scientific
Jump to:
About this item
Durable containers for chilling temperature-sensitive samples.
- Made from polyurethane with metal inserts for stability
- Close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob
- Tapered design for easy stacking
- Precise pour spout and large handles
- The lid and bottom of the bucket/pan are keyed to fit together
- For low-temperature use to −80 °C
Suitable for low temperature use. Ideal for ice, ice slurries and CO₂ pellets.
3 Options Available Below
Product Details & Documents
Durable containers for chilling temperature-sensitive samples.
- Made from polyurethane with metal inserts for stability
- Close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob
- Tapered design for easy stacking
- Precise pour spout and large handles
- The lid and bottom of the bucket/pan are keyed to fit together
- For low-temperature use to −80 °C
Suitable for low temperature use. Ideal for ice, ice slurries and CO₂ pellets.
Product Family Options
Product Information
- CapacityColorAvailabilityPrice
- 490007-646BucketBucket4 LBlackEach$270.90Case of 6$1,625.38Supplier Number: HS28714K
- 490007-648PanPan1 LGreenEach$92.32Case of 6$553.90Supplier Number: HS28721G
- 490007-650PanPan9 LRedEach$246.30Case of 6$1,477.78Supplier Number: HS28729R