DenLine Disposable Lab Coats
: Avantor
Designed to assist researchers who have potential for splash and spray of blood, body fluids or chemicals and are subject to OSHA directive(s) 29 CFR 1910 and OSHA enforcement procedure(s) CPL 02-02.069.
- Latex-free coats are 44" in length
- Polypropylene fabric offers moderate fluid resistance
- SMS (Spunbond-Meltdown-Spunbond) fabric offers high fluid resistance
- Meets OSHA directive(s) 29 CFR 1910 and OSHA enforcement procedure(s) CPL 02-02.069
- Coats feature three pockets, cuffs, and heavy duty snaps
- Disinfect via hot water wash (160˚F)
Fabric of DL3460 series coats is Polypropylene and for DL4260 series it is SMS.
Frequently Bought Together