Diversified Biotech Biodegradable Reagent Reservoirs
Supplier: Diversified Biotech
Certificates
About this item
Disposable, Sterile, and Safe for the Environment.
- Biodegradable- Using a special additive to PET, our reservoirs will safely degrade over time
- Packaging 100% Eco-Friendly- bags are biodegradable & cardboard cases are recyclable
- Unique- "Trough within a trough" maximizes reagent recovery
- Pour-off spouts on all four corners
- Stable- A wide base and reinforced sidewalls avoids spills and provides stability
- Graduated marks on inside wall
- Sterile- Sterilized by gamma irradiation
Frequently Bought Together