GeneMate SnapStrip® PCR Tubes with Individual Attached Dome Caps, 8-Strip 0.2 ml, Scientific Specialties
Supplier: GeneMate
Certificates
About this item
The SnapStrip is an attached 8-strip of tubes with each tube in the strip having its own individual attached dome cap. Another unique and innovative feature of the tubes are user's ability to create a strip of less than 8 tubes with caps by simply breaking the strip where desired.
- Ulta-thin wall for optimum heat transfer
- Certified free from DNase, RNase, and detectable DNA
A strip of 2 or more tubes is easier and safer to use and manipulate than are single tubes with caps.
Flexibility: create a 4-strip today, create a 6-strip tomorrow. The individual tubes may also be used with the 0.2 or 0.5 ml adapters.
Packaging: Packaged as 120 Strips of 8, case has 10 packs of 120 strips of 8.
Frequently Bought Together