ContactUsLinkComponent
GeneMate PCR Tubes with Attached Caps, 8-Strip 0.2 ml, Scientific Specialties
GeneMate PCR Tubes with Attached Caps, 8-Strip 0.2 ml, Scientific Specialties
Catalog # 490003-672
Supplier:  GeneMate
undefined
GeneMate PCR Tubes with Attached Caps, 8-Strip 0.2 ml, Scientific Specialties
Catalog # 490003-672
Supplier:  GeneMate
Specifications
  • Capacity:
    0.2 ml
  • Color:
    Natural
  • Description:
    8-Strip Tubes with Caps
  • Cat. No.:
    490003-672
  • Packaging:
    125 Strips of 8
  • Supplier no.:
    3230-00-210IS
Specifications
About this item
This system virtually ensures the elimination of a major source of contaminationcaps being placed on other than their original tube strips after cycling.
  • Caps need not be replaced due to confusion as to their original orientation on the tubes, accidental dropping, or simply being misplaced
Ordering and inventorying is simplified no more calculating quantities of cap strips to tube strips.
Ordering information: Minimum order requirement of 3 cases.