Serological Pipettes, Class A
Supplier: Heathrow Scientific
Certificates
About this item
Serological pipettes are made out of high-quality borosilicate glass. They also have graduation markings and color-coded bands that are permanently enameled to the glass which allows for quick identification of different volume ranges.
- Wide range of sizes to choose from
- Reusable
- Calibrated to deliver
Serological pipettes are meticulously manufactured for tolerances and accuracy in order to ensure pinpoint precision for all of liquid handling needs.
Certifications: Meets ASTM E-1044 Style 1, Class A specifications
Frequently Bought Together