- Volume:100 µl
- Packed:96/Rack
- Cat. No.:490008-532
- Filter:+
- Sterile:+
- Tip style:Graduated Filtered Pipette Tips
- Supplier no.:F108-96RS-100
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product
- Natural, sterile
- Metal-free tips: All materials used in the manufacture of these tips are free of trace metals
- Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. First-Class Filter Tips: Specially designed for PCR work, these pipette tips prevent airborne contaminants from coming in contact with the barrel of the pipettor. Graduated Tips: Provides reference lines to allow a quick check of pipetting procedures by indicating nominal liquid volume in the tip. For use with but not limited to: Titertek®/Labsystems, Biohit Proline®, Costar® Onepette, Jencons Sealpette®, Labpette™, Oxford-Benchmate®, and GeneMate®.
A mold release agent is NOT used in the manufacturing of these pipette tips. A trace metal certificate is available upon request. Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free.)