Dilux™ Dilution Reservoir
Supplier: DILUX LLC BE
Certificates
About this item
Designed for making serial dilutions of biological samples and chemical reagents using multichannel pipettors, the 12-chamber reservoir holds up to 5 mL of sample
The rounded sides and bottom of the chambers permit complete sample withdrawal. A numerical code inscribed for each chamber ensures accurate sample reference. The fitted lid minimizes sample evaporation and contamination during incubation or storage, while the broad base of the container ensures bench stability.
Frequently Bought Together