ExpellPlus™ Filter Tips
Catalog # 490005-252
Supplier: OXFORD LAB PRODUCTS INC
Specifications
- Volume:10 µl
- Packed:Hinged Racks
- Cat. No.:490005-252
- Filter:+
- Sterile:+
- Tip style:ExpellPlus™ Filter Tips
- Supplier no.:5030030C
Specifications
About this item
The Expell tips without the plus low retention properties are of the same high manufacturing standards, maintaining strict tolerances.
- No Detectable Nucleic acid contamination
- No Detectable PCR Inhibitors
- No Detectable Endonucleases
- No Detectable Endotoxin
- Sterile
Ideal wetting properties and high transparency and purity levels.