Pipettor Tri-Clamp-Ette™
Catalog # 490007-584
Supplier: VWR International
Specifications
- Color:Blue
- Cat. No.:490007-584
- Supplier no.:HS20162A
Specifications
About this item
Conveniently holds three pipettors.
An extension to our Clamp-Ette family of pipetting aids, this innovative product clamps down securely to table tops or benches and holds most major brands of pipettors. The Tri-Clamp-Ette provides the flexibility of housing expensive pipettors practically anywhere in the lab! A screw on each end of the product provides a safe hold with just a few easy turns.