GeneMate Micro Dilution and Storage Tube System, 1.2 ml, Scientific Specialties
Supplier: VWR International
Certificates
About this item
Tube racks feature microtiter footprint and alignment guides for a wide array of consistent, reliable, and automated protocols. Recessed rack bottoms allow for safe and convenient stacking. Polyethylene caps are available for secure and convenient sealing of each tube. Available in 8- or 12-strip format, these caps require minimum insertion pressure to obtain a leak-free seal. Each cap is tabbed for easy handling and removal and caps can easily be snapped apart for single tube handling. RNase-/DNase-/pyrogen-free. 8-strip or 12-strip tube configurations. Bulk bags or 96-place racks. Alphanumeric identification for fast and accurate tube location.
- Serial dilutions using robotics or manual manipulations
- Freezer storage of critical samples
- RIA/EIA/ELISA procedures
- Sample transport
Frequently Bought Together