Orders LinkContactUsLinkComponent
Advansta Buffer Pouches
Advansta Buffer Pouches
Supplier:   Advansta
Advansta Buffer Pouches
Supplier:  Advansta
Certificates
Advansta Buffer Pouches
Advansta Buffer Pouches
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Convenient, pure, fast buffer preparation
  • Quality- Molecular biology-grade buffers of highest purity
  • Convenient- Pre-measured powder in sealed pouches
  • Fast- Dissolve and use right away
  • Standardized- Guaranteed reproducible results for your electrophoresis gels and Western blots
Advansta's Avant buffer pouches are perfect for standardizing your electrophoresis and Western blotting applications. Dissolve a pouch of pre-messured molecular biology-grade chemicals in 500 mL of deionized water to obtain a ready-to-use 1X buffer without any further preparation. Save precious time in the laboratory and generate reproducible results with Avant buffers.
Product Family Options
Product Information
  • Availability
    Price
  • 490005-044
    Buffer PBS Pouches
    Buffer PBS Pouches
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $140.96
    $140.96
    Pack of 20
       $140.96
    Supplier Number: R-01038-020
  • 490005-046
    Buffer TBS Pouches
    Buffer TBS Pouches
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $137.85
    $137.85
    Pack of 20
       $137.85
    Supplier Number: R-01039-020