Order Entry
United States
ContactUsLinkComponent
Shelton Scientific IBI® Horizontal Gel Systems: Multipurpose Design,10×15 cm
Shelton Scientific IBI® Horizontal Gel Systems: Multipurpose Design,10×15 cm
Catalog # 490007-700
Supplier:  IBI Scientific
undefined
Shelton Scientific IBI® Horizontal Gel Systems: Multipurpose Design,10×15 cm
Catalog # 490007-700
Supplier:  IBI Scientific
Supplier Number:  IB53000
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Multipurpose Horizontal Gel System
  • Environmentally Preferable:
  • Cat. No.:
    490007-700
  • Supplier no.:
    IB53000

Specifications

About this item

This 10 x 15 cm Horizontal Gel System is an ideal multi-purpose gel box for both extended high resolution linear runs and economical quick screens requiring buffer recirculation. All models include buffer chamber, safety cover, power cords, casting system with tray, two combs, leveling bubble, and manual.

  • Buffer chamber
  • Safety cover and power cords
  • Casting system with tray
  • Two 2.0 mm thick 16-tooth combs
  • Leveling bubble and manual