GlacierBrand™ Ice Pans, Diversified Biotech
These ice pans are manufactured using durable triple density EPS (polystyrene), for an inexpensive, incredibly lightweight yet rigid ice pan that exhibits superior insulating capabilities.
- Supports ice, dry ice, alcohol/ice slurries, and liquid nitrogen
- Lightweight, yet rigid
- Triple density (EPS)
Supported media include ice, dry ice, alcohol/ice slurries, and liquid nitrogen.
Frequently Bought Together