Order Entry
United States
ContactUsLinkComponent
SP Bel-Art Holdfast Graduated Cylinders, Bel-Art Products, a part of SP
SP Bel-Art Holdfast Graduated Cylinders, Bel-Art Products, a part of SP
Catalog # 490012-188
SP Bel-Art Holdfast Graduated Cylinders, Bel-Art Products, a part of SP
Catalog # 490012-188
Supplier Number:  F28461-1000

Specifications

  • Capacity:
    1000 ml
  • Description:
    Holdfast Graduated Cylinders
  • Form:
    Tall
  • Cat. No.:
    490012-188
  • Supplier no.:
    F28461-1000

Specifications

About this item

Reduce fatigue when repetitive measuring and pouring is required.

  • Graduated markings
  • Easy to read for measuring
  • Convenient handle for pouring
  • Steam autoclavable at 121°C (250°F)

The large handle provides a secure grip for safer, more controlled and convenient pouring. A double scale is accurate and easy to read. The wide hexagonal base prevents tip over and rolling on the benchtop. Hand opening: 89 x 29 mm (3 1/2 x 1 1/8").