PVDF Pure Transfer Membranes, GVS Filter Technology
Supplier: GVS FILTER TECHNOLOGY
Certificates
About this item
PVDF transfer membranes.
- High binding capacity, which prevents protein from passing through membrane
- Low background, excellent signal to noise ratio
- Broad chemical compatibility
GVS PVDF Pure is naturally hydrophobic, available with 0.45 μm pore size for high retention blotting.
Frequently Bought Together