NitroPlus Transfer Membranes, GVS Filter Technology
Supplier: GVS FILTER TECHNOLOGY
Certificates
About this item
Supported Nitrocellulose (MCE) membranes.
- Supported for rigorous handling and multiple reprobings
- Strong construction will not curl, bend, or crack after baking
- High sensitivity, low background
- BSA binding capacity up to 100 ug/cm2
- Triton-free
GVS Filter Technology NitroPlus Supported Transfer Membranes combine the binding characteristics of a nitrocellulose membrane with the strength of a nylon membrane. NitroPure can easily be used in any protocol utilizing unsupported nitrocellulose transfer membranes.
Frequently Bought Together