NitroPure Transfer Membranes, GVS Filter Technology
Supplier: GVS FILTER TECHNOLOGY
Certificates
About this item
Nitrocellulose membranes.
- High sensitivity ensures excellent results in all transfers
- Low background
- Easily blocked
- BSA binding capacity up to 100 µm/cm2
- Optimum resolution
GVS Life Sciences Nitrocellulose Transfer Membranes are the membrane of choice for all protein or immunoblotting applications.
Frequently Bought Together