Gel Drying Frames, Diversified Biotech
About this item
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers
The 24 x 24 cm frames allow drying of larger format gels, while the 14 x 14 cm frames work well with minigels and most other formats. Packaged with 2 open frames, 2 solid back plates, 8 side clips, and 50 sheets of pre-cut cellophane.
4 Options Available Below
Product Details & Documents
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers
The 24 x 24 cm frames allow drying of larger format gels, while the 14 x 14 cm frames work well with minigels and most other formats. Packaged with 2 open frames, 2 solid back plates, 8 side clips, and 50 sheets of pre-cut cellophane.
Product Family Options
Product Information
- AvailabilityPrice
- 490006-662Gel Drying Frames, 14 x 14 cmSupplier Number: AB-110760
- 490006-664Gel Drying Frames, 24 x 24 cmSupplier Number: AB-110761
- 490006-66650 Sheets Cellophane for 14 x 14 cm FrameSupplier Number: AB-110762
- 490006-66850 Sheets Cellophane for 24 x 24 cm FrameSupplier Number: AB-110763