Ward's® Single Probes Sound Sensor
Catalog # 470014-848
Supplier: Wards
Specifications
- Description:Sound Probe
- Cat. No.:470014-848
- Supplier no.:WSP-108-500
Specifications
About this item
Explore the science of sound while measuring both intensity and waves with the Ward's Single Probes Sound Sensor.
- Measure intensity of sound waves
- Two operational modes
- Free activity guide
Two operational modes make it easy to measure both sound-pressure (decibels) and frequency. Sensor includes free activity guide with activities designed for use with each probe, available digitally online after purchase. Range: 40 – 110 dB; Resolution: 0.1 dB. Requires Ward's Single Probes Base Unit, available separately.