Order Entry
United States
ContactUsLinkComponent
Colloidal Iron Histology Stain Kit, Diagnostic Biosystems
Colloidal Iron Histology Stain Kit, Diagnostic Biosystems
Catalog # 95027-178
CAS Number:  
undefined
Colloidal Iron Histology Stain Kit, Diagnostic Biosystems
Catalog # 95027-178
Supplier Number:  KT007
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Colloidal Iron Histology Stain Kit
  • Cat. No.:
    95027-178
  • No. of reactions:
    100
  • Supplier no.:
    KT 007

Specifications

About this item

The Colloidal Iron Stain Kit is designed for the histological visualization of acid mucopolysaccharides.