Order Entry
United States
ContactUsLinkComponent
Alcian Blue Histology Stain Kit, pH 1.0, Diagnostic Biosystems
Alcian Blue Histology Stain Kit, pH 1.0, Diagnostic Biosystems
Catalog # 95027-172
CAS Number:  
undefined
Alcian Blue Histology Stain Kit, pH 1.0, Diagnostic Biosystems
Catalog # 95027-172
Supplier Number:  KT002
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Alcian Blue Histology Stain Kit, pH 1.0
  • Cat. No.:
    95027-172
  • No. of reactions:
    100
  • Supplier no.:
    KT 002

Specifications

About this item

This kit stains strongly sulfated mucosubstances

Strongly sulfated mucosubstances stain blue in paraffin tissue sections, while the nuclei stain pink to red and the cytoplasm and background stain pale pink.