ContactUsLinkComponent
Omniprobe 3 Post Copper, Electron Microscopy Sciences
Omniprobe 3 Post Copper, Electron Microscopy Sciences
Catalog # 102773-584
Omniprobe 3 Post Copper, Electron Microscopy Sciences
Catalog # 102773-584
Supplier Number:  75964-07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    3 Post Copper with 1 lower egde
  • Cat. No.:
    102773-584
  • Supplier no.:
    75964-07
Specifications
About this item
The 3 post copper lift-out grids include multiple indexed mounting locations with both vertical bars and 'V' shaped attachment surfaces.
Packaging: Packaged in glass vials.