ContactUsLinkComponent
Omniprobe Autoprobe 250 Tungsten Probe Tips, Electron Microscopy Sciences
Omniprobe Autoprobe 250 Tungsten Probe Tips, Electron Microscopy Sciences
  102681-282
undefined
Omniprobe Autoprobe 250 Tungsten Probe Tips, Electron Microscopy Sciences
  102681-282
 :  75960-05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    AP250 Tungsten Probe Tip
  • Cat. No.:
    102681-282
  • Supplier no.:
    75960-05
Custom tip designed from tungsten. Tip radius is < 0.5 µm with 6° taper angle for maximum lifetime service. Compatible with Short-Cut™ system for direct conversion to TEM grid.
 : Xtreme Access Compatible Probe Point Holder is for use with 1/2" tungsten probe tips. Comes with a storage vessel. The AP250 AutoProbe™ tip holder uses the tungsten probe tip. Compatible with the Short-Cut™ for direct conversion to TEM Grid. Comes with a storage vial and handle.