Orders LinkContactUsLinkComponent
Omniprobe Single Stub and Two TEM Grids Holders, Electron Microscopy Sciences
Omniprobe Single Stub and Two TEM Grids Holders, Electron Microscopy Sciences
Omniprobe Single Stub and Two TEM Grids Holders, Electron Microscopy Sciences
Certificates
Omniprobe Single Stub and Two TEM Grids Holders, Electron Microscopy Sciences
Omniprobe Single Stub and Two TEM Grids Holders, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This station is meant for 2 TEM grids and 1 sample pin stub. Available in two versions: Stainless Steel (slightly magnetic), and Aluminum (Al – non-magnetic). Long post - standard is 1/8"(3.2 mm) diameter and 0.32" (8.1 mm) length.
Delivery information: Comes with an aluminum sample stub.
Product Family Options
Product Information
  • Availability
    Price
  • 102681-318
    TEM grid and sample holder, stainless steel
    TEM grid and sample holder, stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $2,207.69
    $2,207.69
    Each
       $2,207.69
    Supplier Number: 75971-SS
  • Discontinued
    Supplier Number: -