Metallurgical Mount Holder, 1-¹/₄" and 30 mm, Electron Microscopy Sciences
This mount holder which is made from machined aluminum with stainless steel Allen screws is specifically for 1 1/4” and 30mm metallurgical mounts.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.