Specifications
- Description:45° Multi Holder for 3 pin stubs,
- Environmentally Preferable:
- Cat. No.:102098-322
- Supplier no.:75952-60
Specifications
About this item
This 25.4 mm in diameter 45 Degree multi pin holder is designed to save time. It allows for a higher SE signal without having to tilt the specimen It is made from machined aluminum with stainless steel Allen set screws. It accommodates up to three standard 12.7 mm (1/2") pin stubs with 3.2 mm (1/8") pin.